Assay Design
Gene Transcripts
- Gene Symbol
- Slc25a23
- Entrez Gene ID
-
66972
- Gene Name
- solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23
- Gene Aliases
-
2310067G05Rik, SCaMC-3
- Chromosome Location
-
Chr.17: 57043711 - 57059880 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_025877.4 |
NP_080153.2 |
6 - 7 |
986 |
76 |
GenBank mRNA |
BC072660.1 |
- |
6 - 7 |
982 |
76 |
View Full Product Details