Assay Design



Gene Transcripts

Gene Symbol
Ccdc38
Entrez Gene ID
237465
Gene Name
coiled-coil domain containing 38
Gene Aliases
4933417K05Rik
Chromosome Location
Chr.10: 93540567 - 93584326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175488.6 NP_780697.2 2 - 3 225 141
GenBank mRNA AK029803.1 - 2 - 3 226 141
AK030198.1 - 2 - 3 214 141

View Full Product Details