Assay Design



Gene Transcripts

Gene Symbol
Ntn4
Entrez Gene ID
57764
Gene Name
netrin 4
Gene Aliases
-
Chromosome Location
Chr.10: 93641049 - 93745972 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021320.3 NP_067295.2 4 - 5 1391 101
GenBank mRNA AF268066.1 - 4 - 5 986 101
AF281278.1 - 4 - 5 1239 101
BC119182.1 - 4 - 5 1073 101
BC119184.1 - 4 - 5 1073 101
CX205916.1 - 2 - 3 202 101

View Full Product Details