Assay Design



Gene Transcripts

Gene Symbol
Ntn4
Entrez Gene ID
57764
Gene Name
netrin 4
Gene Aliases
-
Chromosome Location
Chr.10: 93641049 - 93745972 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021320.3 NP_067295.2 6 - 7 1798 89
GenBank mRNA AF268066.1 - 6 - 7 1393 89
AF281278.1 - 6 - 7 1646 89
BC119182.1 - 6 - 7 1480 89
BC119184.1 - 6 - 7 1480 89
CF537120.1 - 2 - 3 392 89

View Full Product Details