Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145158.3 | NP_660140.1 | 4 - 5 | 2466 | 69 |
GenBank mRNA | AF468645.1 | - | 4 - 5 | 2417 | 69 |
AK154230.1 | - | 4 - 5 | 2436 | 69 | |
AK155037.1 | - | 4 - 5 | 2446 | 69 | |
AK155306.1 | - | 4 - 5 | 2460 | 69 | |
AK155554.1 | - | 4 - 5 | 2462 | 69 | |
AK155797.1 | - | 4 - 5 | 2467 | 69 | |
AK155878.1 | - | 4 - 5 | 2464 | 69 | |
AK155879.1 | - | 4 - 5 | 2460 | 69 | |
AK155909.1 | - | 4 - 5 | 2466 | 69 | |
AK155913.1 | - | 4 - 5 | 2436 | 69 | |
AK155918.1 | - | 4 - 5 | 2493 | 69 | |
AK170288.1 | - | 4 - 5 | 2436 | 69 | |
AK170391.1 | - | 4 - 5 | 2436 | 69 | |
AK170801.1 | - | 4 - 5 | 2467 | 69 | |
AK170822.1 | - | 4 - 5 | 2488 | 69 | |
AK170953.1 | - | 4 - 5 | 2482 | 69 | |
AK171015.1 | - | 4 - 5 | 2488 | 69 | |
AK171020.1 | - | 4 - 5 | 2454 | 69 | |
AK171303.1 | - | 4 - 5 | 2438 | 69 | |
AK171309.1 | - | 4 - 5 | 2463 | 69 | |
AK171579.1 | - | 4 - 5 | 2436 | 69 | |
AK171592.1 | - | 4 - 5 | 2436 | 69 | |
AK171616.1 | - | 4 - 5 | 2436 | 69 | |
BC053753.1 | - | 4 - 5 | 2750 | 69 | |
GQ872457.1 | - | 4 - 5 | 2361 | 69 | |
GQ872460.1 | - | 4 - 5 | 2361 | 69 |