Assay Design



Gene Transcripts

Gene Symbol
Wdr43
Entrez Gene ID
72515
Gene Name
WD repeat domain 43
Gene Aliases
2610318G08Rik, AU020887, AV024208, Wrd43, mKIAA0007
Chromosome Location
Chr.17: 71615900 - 71659031 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175639.1 NP_783570.1 11 - 12 1475 96
GenBank mRNA AK012043.1 - 11 - 12 1460 96
AK032907.1 - 11 - 12 1449 96
AK129031.1 - 11 - 12 1393 96
AK152321.1 - 11 - 12 1465 96
BC079913.1 - 10 - 11 1176 96

View Full Product Details