Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033289.2 | NP_001028461.2 | 11 - 12 | 2226 | 63 |
GenBank mRNA | AK077026.1 | - | 11 - 12 | 2318 | 63 |
AK138779.1 | - | 11 - 12 | 2604 | 63 | |
BC103685.1 | - | 11 - 12 | 2226 | 63 | |
BC104737.1 | - | 11 - 12 | 2226 | 63 | |
BC105676.1 | - | 11 - 12 | 2225 | 63 |