Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033289.2 | NP_001028461.2 | 7 - 8 | 1745 | 93 |
GenBank mRNA | AF139193.1 | - | 2 - 3 | 98 | 93 |
AK077026.1 | - | 7 - 8 | 1837 | 93 | |
AK138779.1 | - | 7 - 8 | 2122 | 93 | |
BC103685.1 | - | 7 - 8 | 1745 | 93 | |
BC104737.1 | - | 7 - 8 | 1745 | 93 | |
BC105676.1 | - | 7 - 8 | 1744 | 93 |