Assay Design



Gene Transcripts

Gene Symbol
Iqcf6
Entrez Gene ID
100041096
Gene Name
IQ motif containing F6
Gene Aliases
-
Chromosome Location
Chr.9: 106626582 - 106627675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101628.1 NP_001095098.1 1 - 2 101 74

View Full Product Details