Assay Design
Gene Transcripts
- Gene Symbol
- Cylc1
- Entrez Gene ID
-
67407
- Gene Name
- cylicin, basic protein of sperm head cytoskeleton 1
- Gene Aliases
-
4930401M13Rik
- Chromosome Location
-
Chr.X: 111110418 - 111125961 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026134.2 |
NP_080410.2 |
3 - 4 |
154 |
112 |
GenBank mRNA |
AK015303.1 |
- |
3 - 4 |
156 |
112 |
View Full Product Details