Assay Design



Gene Transcripts

Gene Symbol
Peli3
Entrez Gene ID
240518
Gene Name
pellino 3
Gene Aliases
6030441F14Rik, A930011L17, BC028931
Chromosome Location
Chr.19: 4929717 - 4943127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172835.3 NP_766423.3 4 - 5 543 87
GenBank mRNA AK044418.1 - 4 - 5 490 87
BC028931.1 - 4 - 5 501 87

View Full Product Details