Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007903.4 | NP_031929.1 | 1 - 2 | 261 | 113 |
GenBank mRNA | AK048006.1 | - | 1 - 2 | 184 | 113 |
AK049775.1 | - | 1 - 2 | 247 | 113 | |
AK079150.1 | - | 1 - 2 | 260 | 113 | |
BC137727.1 | - | 1 - 2 | 197 | 113 | |
BC137730.1 | - | 1 - 2 | 197 | 113 | |
U32330.1 | - | 1 - 2 | 401 | 113 |