Assay Design
Gene Transcripts
- Gene Symbol
- Sun5
- Entrez Gene ID
-
76407
- Gene Name
- Sad1 and UNC84 domain containing 5
- Gene Aliases
-
1700021O15Rik, SPAG4L-2, SRG4, Spag4l
- Chromosome Location
-
Chr.2: 153856188 - 153871095 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK006225.1 |
- |
4 - 5 |
327 |
72 |
AY307077.1 |
- |
4 - 5 |
346 |
72 |
FJ667498.1 |
- |
5 - 6 |
335 |
72 |
View Full Product Details