Assay Design



Gene Transcripts

Gene Symbol
Rilp
Entrez Gene ID
280408
Gene Name
Rab interacting lysosomal protein
Gene Aliases
AI835553, Gm857
Chromosome Location
Chr.11: 75510094 - 75513168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029938.2 NP_001025109.1 4 - 5 663 54
GenBank mRNA AK139016.1 - 4 - 5 655 54
AK160517.1 - 2 - 3 456 54
BC051502.1 - 1 - 2 122 54
BC116750.1 - 4 - 5 647 54
BC119166.1 - 4 - 5 647 54

View Full Product Details