Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172150.4 | NP_742162.2 | 11 - 12 | 1085 | 53 |
GenBank mRNA | AK088467.1 | - | 11 - 12 | 1086 | 53 |
AK144188.1 | - | 11 - 12 | 1185 | 53 | |
AK167194.1 | - | 11 - 12 | 1101 | 53 | |
BC037708.1 | - | 11 - 12 | 1054 | 53 |