Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153195.2 | NP_694875.2 | 6 - 7 | 1059 | 59 |
GenBank mRNA | AK010499.1 | - | 4 - 5 | 337 | 59 |
AK147716.1 | - | - | 359 | 59 | |
AK147797.1 | - | 6 - 7 | 1011 | 59 | |
AK147907.1 | - | 6 - 7 | 978 | 59 | |
AK150190.1 | - | 6 - 7 | 834 | 59 | |
AK150626.1 | - | 6 - 7 | 1017 | 59 | |
AK151025.1 | - | 6 - 7 | 1017 | 59 | |
AK152513.1 | - | 6 - 7 | 1017 | 59 | |
BC032153.1 | - | 6 - 7 | 1001 | 59 |