Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021441.2 | NP_067416.2 | 4 - 5 | 427 | 66 |
GenBank mRNA | AK030522.1 | - | 4 - 5 | 394 | 66 |
AK052250.1 | - | 4 - 5 | 394 | 66 | |
AK087534.1 | - | 4 - 5 | 417 | 66 | |
AK154846.1 | - | 4 - 5 | 411 | 66 | |
AK156718.1 | - | 4 - 5 | 394 | 66 | |
BC051944.1 | - | 4 - 5 | 406 | 66 | |
BC058383.1 | - | 4 - 5 | 369 | 66 | |
Y09974.1 | - | 4 - 5 | 554 | 66 |