Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113180.1 | NP_001106651.1 | 16 - 17 | 3023 | 103 |
NM_019691.4 | NP_062665.3 | 16 - 17 | 3023 | 103 | |
GenBank mRNA | AB022913.1 | - | 16 - 17 | 2757 | 103 |
AB079073.1 | - | 15 - 16 | 2544 | 103 | |
AB102777.1 | - | 15 - 16 | 2544 | 103 | |
AB104708.1 | - | 4 - 5 | 792 | 103 | |
AK036193.1 | - | 5 - 6 | 906 | 103 | |
AK135034.1 | - | 16 - 17 | 2731 | 103 |