Assay Design
Gene Transcripts
- Gene Symbol
- Sec14l1
- Entrez Gene ID
-
74136
- Gene Name
- SEC14-like 1 (S. cerevisiae)
- Gene Aliases
-
1200017E04Rik, 2810012L19Rik, Naa-35
- Chromosome Location
-
Chr.11: 117115172 - 117159268 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_028777.3 |
NP_083053.2 |
16 - 17 |
2357 |
60 |
GenBank mRNA |
AK004818.1 |
- |
16 - 17 |
2282 |
60 |
View Full Product Details