Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166506.1 | NP_001159978.1 | 1 - 2 | 186 | 131 |
NM_001166507.1 | NP_001159979.1 | 1 - 2 | 186 | 131 | |
NM_028777.3 | NP_083053.2 | 1 - 2 | 186 | 131 | |
GenBank mRNA | AK004818.1 | - | 1 - 2 | 111 | 131 |
AK047396.1 | - | 1 - 2 | 191 | 131 | |
BB646963.1 | - | 1 - 2 | 186 | 131 | |
BC005766.1 | - | 1 - 2 | 144 | 131 | |
BC086620.1 | - | 1 - 2 | 120 | 131 |