Assay Design



Gene Transcripts

Gene Symbol
Rap1gap2
Entrez Gene ID
380711
Gene Name
RAP1 GTPase activating protein 2
Gene Aliases
AU067654, Garnl4, Gm1561, mKIAA1039
Chromosome Location
Chr.11: 74383483 - 74610910 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015046.2 NP_001015046.1 16 - 17 1516 87
GenBank mRNA AK038834.1 - 6 - 7 579 87
AK122424.1 - 2 - 3 309 87
AK147290.1 - 17 - 18 1498 87
BC107388.2 - 7 - 8 722 87

View Full Product Details