Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128086.2 | NP_001121558.2 | 20 - 21 | 3346 | 124 |
NM_001177396.1 | NP_001170867.1 | 7 - 8 | 1249 | 124 | |
NM_001177397.1 | NP_001170868.1 | 6 - 7 | 792 | 124 | |
NM_001177398.1 | NP_001170869.1 | 6 - 7 | 1072 | 124 | |
NM_181398.3 | NP_852063.1 | 5 - 6 | 997 | 124 | |
GenBank mRNA | AK032061.1 | - | 4 - 5 | 448 | 124 |
AK032211.1 | - | 6 - 7 | 1074 | 124 | |
AK034154.1 | - | 7 - 8 | 812 | 124 | |
AK038609.1 | - | 4 - 5 | 676 | 124 | |
AK049899.1 | - | 5 - 6 | 942 | 124 | |
AK132228.1 | - | 7 - 8 | 1216 | 124 | |
AK163750.1 | - | 5 - 6 | 971 | 124 | |
BC098373.1 | - | 6 - 7 | 792 | 124 |