Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128086.2 | NP_001121558.2 | 23 - 24 | 3510 | 104 |
NM_001177396.1 | NP_001170867.1 | 10 - 11 | 1413 | 104 | |
NM_001177397.1 | NP_001170868.1 | 9 - 10 | 956 | 104 | |
NM_001177398.1 | NP_001170869.1 | 9 - 10 | 1236 | 104 | |
NM_181398.3 | NP_852063.1 | 8 - 9 | 1161 | 104 | |
GenBank mRNA | AK032061.1 | - | 7 - 8 | 612 | 104 |
AK032211.1 | - | 9 - 10 | 1238 | 104 | |
AK034154.1 | - | 10 - 11 | 976 | 104 | |
AK038609.1 | - | 7 - 8 | 840 | 104 | |
AK049899.1 | - | 8 - 9 | 1106 | 104 | |
AK132228.1 | - | 10 - 11 | 1380 | 104 | |
AK163750.1 | - | 8 - 9 | 1135 | 104 | |
BC098373.1 | - | 9 - 10 | 956 | 104 |