Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159324.1 | NP_001152796.1 | 15 - 16 | 2360 | 88 |
NM_008064.3 | NP_032090.3 | 15 - 16 | 2450 | 88 | |
GenBank mRNA | AK052211.1 | - | 15 - 16 | 2431 | 88 |
AK088481.1 | - | 15 - 16 | 2426 | 88 | |
AK139333.1 | - | 15 - 16 | 2398 | 88 | |
AK141546.1 | - | 15 - 16 | 2433 | 88 | |
AK145631.1 | - | 14 - 15 | 1866 | 88 | |
AK146538.1 | - | 15 - 16 | 2354 | 88 | |
AK150970.1 | - | 15 - 16 | 2408 | 88 | |
BC010210.1 | - | 15 - 16 | 2414 | 88 |