Assay Design



Gene Transcripts

Gene Symbol
Zmynd8
Entrez Gene ID
228880
Gene Name
zinc finger, MYND-type containing 8
Gene Aliases
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7
Chromosome Location
Chr.2: 165784152 - 165898984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291158.1 NP_001278087.1 16 - 17 4857 84
NM_027230.5 NP_081506.3 16 - 17 2935 84
GenBank mRNA AK014397.1 - 3 - 4 992 84
BC041797.1 - 5 - 6 1663 84
BC048186.1 - 16 - 17 2742 84

View Full Product Details