Assay Design



Gene Transcripts

Gene Symbol
Zmynd8
Entrez Gene ID
228880
Gene Name
zinc finger, MYND-type containing 8
Gene Aliases
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7
Chromosome Location
Chr.2: 165784152 - 165898984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252584.2 NP_001239513.1 16 - 17 2879 68
NM_001252585.1 NP_001239514.1 15 - 16 2771 68
NM_001281926.1 NP_001268855.1 16 - 17 2778 68
NM_001291158.1 NP_001278087.1 17 - 18 5044 68
NM_027230.5 NP_081506.3 17 - 18 3122 68
GenBank mRNA AB093284.2 - 17 - 18 5036 68
AK014397.1 - 4 - 5 1179 68
AK132427.1 - 15 - 16 2773 68
AK147620.1 - 16 - 17 2779 68
AK155864.1 - 16 - 17 2684 68
BC041797.1 - 6 - 7 1850 68
BC048186.1 - 17 - 18 2929 68

View Full Product Details