Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037723.3 | NP_001032812.2 | 27 - 28 | 3741 | 61 |
NM_001037724.4 | NP_001032813.1 | 27 - 28 | 3677 | 61 | |
NM_001109756.1 | NP_001103226.1 | 27 - 28 | 3645 | 61 | |
NM_007406.2 | NP_031432.2 | 27 - 28 | 3829 | 61 | |
GenBank mRNA | AK138911.1 | - | 27 - 28 | 3661 | 61 |
AK154262.1 | - | 27 - 28 | 3732 | 61 | |
AK154652.1 | - | 27 - 28 | 3647 | 61 | |
AK155646.1 | - | 27 - 28 | 3646 | 61 | |
AK155831.1 | - | 27 - 28 | 3648 | 61 | |
BC057682.1 | - | 17 - 18 | 1972 | 61 | |
BC115833.1 | - | 25 - 26 | 3523 | 61 | |
U12919.1 | - | 27 - 28 | 3830 | 61 |