Assay Design



Gene Transcripts

Gene Symbol
Adcy7
Entrez Gene ID
11513
Gene Name
adenylate cyclase 7
Gene Aliases
AA407758
Chromosome Location
Chr.8: 88272367 - 88329965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037723.3 NP_001032812.2 5 - 6 965 74
NM_001037724.4 NP_001032813.1 5 - 6 901 74
NM_001109756.1 NP_001103226.1 5 - 6 869 74
NM_007406.2 NP_031432.2 5 - 6 1053 74
GenBank mRNA AK138911.1 - 5 - 6 885 74
AK154262.1 - 5 - 6 956 74
AK154652.1 - 5 - 6 871 74
AK155646.1 - 5 - 6 870 74
AK155831.1 - 5 - 6 872 74
BC115833.1 - 3 - 4 747 74
U12919.1 - 5 - 6 1054 74

View Full Product Details