Assay Design



Gene Transcripts

Gene Symbol
Trp53tg5
Entrez Gene ID
73603
Gene Name
transformation related protein 53 target 5
Gene Aliases
1700126L10Rik
Chromosome Location
Chr.2: 164467903 - 164478812 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271575.1 NP_001258504.1 4 - 5 816 85
GenBank mRNA AK007294.1 - 4 - 5 817 85

View Full Product Details