Assay Design



Gene Transcripts

Gene Symbol
Pigt
Entrez Gene ID
78928
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class T
Gene Aliases
2510012P17Rik, 4930534E15Rik, CGI-06, NDAP, Ndap7
Chromosome Location
Chr.2: 164497511 - 164508301 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133779.2 NP_598540.2 1 - 2 230 67
GenBank mRNA AK019717.2 - 1 - 2 221 67
AK044499.1 - 1 - 2 214 67
AK049990.1 - 1 - 2 223 67
AK090020.1 - 1 - 2 223 67
AK153462.1 - 1 - 2 216 67
AK153464.1 - 1 - 2 216 67
AK155057.1 - 1 - 2 217 67
AK157226.1 - 1 - 2 213 67
AK162573.1 - 1 - 2 210 67
BC016520.1 - 1 - 2 194 67
BC034175.1 - 1 - 2 206 67
BU058874.1 - 1 - 2 230 67

View Full Product Details