Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013865.2 | NP_038893.1 | 11 - 12 | 916 | 89 |
NM_180956.1 | NP_851287.1 | 11 - 12 | 916 | 89 | |
GenBank mRNA | AB033922.1 | - | 11 - 12 | 830 | 89 |
AK004689.1 | - | 11 - 12 | 862 | 89 | |
AK037487.1 | - | 11 - 12 | 842 | 89 | |
AK154927.1 | - | 11 - 12 | 883 | 89 | |
AK185967.1 | - | 1 - 2 | 56 | 89 | |
AK188494.1 | - | 1 - 2 | 56 | 89 | |
BC018504.1 | - | 11 - 12 | 850 | 89 |