Assay Design



Gene Transcripts

Gene Symbol
Ctcfl
Entrez Gene ID
664799
Gene Name
CCCTC-binding factor (zinc finger protein)-like
Gene Aliases
Boris, OTTMUSG00000016680
Chromosome Location
Chr.2: 173093609 - 173119525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081387.2 NP_001074856.1 8 - 9 1738 55
GenBank mRNA BC151026.1 - 8 - 9 1613 55
DQ153171.1 - 8 - 9 1738 55

View Full Product Details