Assay Design



Gene Transcripts

Gene Symbol
Taok1
Entrez Gene ID
216965
Gene Name
TAO kinase 1
Gene Aliases
2810468K05Rik, AU020252, D130018F14Rik, Map3k16, Markk, Psk2, mKIAA1361
Chromosome Location
Chr.11: 77529162 - 77607860 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144825.2 NP_659074.2 15 - 16 2399 151
GenBank mRNA BC016522.1 - 3 - 4 393 151
BC047271.1 - 14 - 15 1746 151
BC151012.1 - 15 - 16 2193 151
BC151015.1 - 15 - 16 2193 151
BC151016.1 - 15 - 16 2192 151
CN717576.1 - 2 - 3 207 151
CN724388.1 - 3 - 4 492 151

View Full Product Details