Assay Design



Gene Transcripts

Gene Symbol
Cdk17
Entrez Gene ID
237459
Gene Name
cyclin-dependent kinase 17
Gene Aliases
6430598J10Rik, Pctk2
Chromosome Location
Chr.10: 93160876 - 93250688 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146239.2 NP_666351.2 1 - 2 327 140
GenBank mRNA AK078325.1 - 1 - 2 328 140
BC064815.1 - 1 - 2 312 139
CJ117086.1 - 1 - 2 327 140

View Full Product Details