Assay Design



Gene Transcripts

Gene Symbol
Ces5a
Entrez Gene ID
67935
Gene Name
carboxylesterase 5A
Gene Aliases
1700081L16Rik, 1700122C07Rik, BB081581, Ces7, Gm503, cauxin
Chromosome Location
Chr.8: 93499063 - 93535881 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003951.2 NP_001003951.1 11 - 12 1430 64
GenBank mRNA AB186393.1 - 11 - 12 1430 64

View Full Product Details