Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025481.2 | NP_079757.2 | 1 - 2 | 532 | 88 |
GenBank mRNA | AY685230.1 | - | 1 - 2 | 212 | 88 |
BC138786.1 | - | 2 - 3 | 97 | 88 | |
BC138788.1 | - | 2 - 3 | 108 | 88 | |
BU058366.1 | - | 1 - 2 | 60 | 88 |