Assay Design



Gene Transcripts

Gene Symbol
Sipa1l2
Entrez Gene ID
244668
Gene Name
signal-induced proliferation-associated 1 like 2
Gene Aliases
BC058408, mKIAA1389
Chromosome Location
Chr.8: 125418063 - 125569863 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081337.1 NP_001074806.1 11 - 12 3656 59
GenBank mRNA AK028440.1 - 11 - 12 2770 59
AK122501.1 - 9 - 10 1933 59
BC058408.1 - 2 - 3 129 59
BC072593.1 - 13 - 14 3942 59
BC089367.1 - 1 - 2 42 59
BC150682.1 - 11 - 12 3701 59

View Full Product Details