Assay Design



Gene Transcripts

Gene Symbol
Atp2c2
Entrez Gene ID
69047
Gene Name
ATPase, Ca++ transporting, type 2C, member 2
Gene Aliases
1810010G06Rik, Spca2, mKIAA0703
Chromosome Location
Chr.8: 119700009 - 119757718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026922.1 NP_081198.1 17 - 18 1807 97
GenBank mRNA AK220351.1 - 15 - 16 1392 97
BC026636.1 - 1 - 2 125 97
BC042772.1 - 8 - 9 825 97
EF688288.1 - 17 - 18 1651 97

View Full Product Details