Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029993.1 | NP_001025164.1 | 1 - 2 | 180 | 113 |
NM_001029994.1 | NP_001025165.1 | 1 - 2 | 180 | 113 | |
GenBank mRNA | AK017933.1 | - | 1 - 2 | 157 | 113 |
AK031413.1 | - | 1 - 2 | 129 | 113 | |
AK159881.1 | - | 1 - 2 | 116 | 113 | |
AK166802.1 | - | 1 - 2 | 135 | 113 | |
BC030495.1 | - | 1 - 2 | 160 | 113 | |
BY235998.1 | - | 1 - 2 | 187 | 113 | |
CD348551.1 | - | 1 - 2 | 149 | 113 |