Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135100.2 | NP_001128572.1 | 3 - 4 | 739 | 83 |
NM_029646.3 | NP_083922.1 | 3 - 4 | 739 | 83 | |
GenBank mRNA | AK008082.1 | - | 3 - 4 | 687 | 83 |
BC016254.1 | - | 3 - 4 | 718 | 83 | |
EU636995.1 | - | 2 - 3 | 156 | 83 | |
EU636996.1 | - | 2 - 3 | 156 | 83 |