Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286006.1 | NP_001272935.1 | 14 - 15 | 2054 | 61 |
NM_172569.4 | NP_766157.1 | 14 - 15 | 2093 | 61 | |
GenBank mRNA | AK046395.1 | - | 14 - 15 | 2088 | 61 |
AK077635.1 | - | 14 - 15 | 2095 | 61 | |
AK078095.1 | - | 14 - 15 | 2045 | 61 | |
AK129438.1 | - | 14 - 15 | 2071 | 61 | |
AK134361.1 | - | 14 - 15 | 2174 | 61 | |
BC003195.1 | - | 11 - 12 | 1539 | 61 | |
BC005545.1 | - | 1 - 2 | 100 | 61 | |
BC033048.1 | - | 2 - 3 | 293 | 61 | |
BC039964.1 | - | 14 - 15 | 2055 | 61 | |
BC054452.1 | - | 14 - 15 | 2030 | 61 |