Assay Design



Gene Transcripts

Gene Symbol
Unk
Entrez Gene ID
217331
Gene Name
unkempt homolog (Drosophila)
Gene Aliases
B230379M23Rik, Zc3h5, Zc3hdc5, mKIAA1753
Chromosome Location
Chr.11: 116030295 - 116061214 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286006.1 NP_001272935.1 7 - 8 1034 108
NM_172569.4 NP_766157.1 7 - 8 1034 108
GenBank mRNA AK046395.1 - 7 - 8 1029 108
AK077635.1 - 7 - 8 1036 108
AK078095.1 - 7 - 8 1025 108
AK129438.1 - 7 - 8 1012 108
AK134361.1 - 7 - 8 1154 108
BC003195.1 - 4 - 5 480 108
BC039964.1 - 7 - 8 996 108
BC054452.1 - 7 - 8 971 108

View Full Product Details