Assay Design



Gene Transcripts

Gene Symbol
Spg7
Entrez Gene ID
234847
Gene Name
spastic paraplegia 7 homolog (human)
Gene Aliases
AI452278, AU015315, Cmar, PGN
Chromosome Location
Chr.8: 123065508 - 123097751 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153176.4 NP_694816.3 5 - 6 775 121
GenBank mRNA AF512565.1 - 5 - 6 807 121
AF547215.1 - 5 - 6 1104 121
AK145540.1 - 5 - 6 776 121
AK171647.1 - 4 - 5 674 121
BC024986.1 - 3 - 4 430 121
BC051051.1 - 5 - 6 709 121
BC055488.1 - - 620 121
BC138141.1 - 5 - 6 1002 121

View Full Product Details