Assay Design



Gene Transcripts

Gene Symbol
Tango6
Entrez Gene ID
272538
Gene Name
transport and golgi organization 6
Gene Aliases
AW413431, E330010G16, Tmco7
Chromosome Location
Chr.8: 106683068 - 106851439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173037.1 NP_766625.1 7 - 8 1474 67
GenBank mRNA AK047206.1 - 2 - 3 192 67
AK054283.1 - 7 - 8 1428 67
AK077757.1 - 7 - 8 1459 67
AK085030.1 - 7 - 8 1474 67
AK173250.1 - 7 - 8 1381 67
BC066134.1 - 7 - 8 1404 67
BC066869.1 - 7 - 8 1404 67
BC069838.1 - 7 - 8 1404 67

View Full Product Details