Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177150.2 | NP_796124.1 | 12 - 13 | 1444 | 69 |
GenBank mRNA | AK089172.1 | - | 12 - 13 | 1493 | 69 |
AK090340.1 | - | 12 - 13 | 1445 | 69 | |
AK143613.1 | - | 12 - 13 | 1407 | 69 | |
AK164039.1 | - | 8 - 9 | 1045 | 69 | |
AK167376.1 | - | 12 - 13 | 1445 | 69 | |
BC022690.1 | - | 3 - 4 | 304 | 69 | |
BC121824.1 | - | 12 - 13 | 1445 | 69 |