Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291076.1 | NP_001278005.1 | 4 - 5 | 455 | 78 |
NM_198214.3 | NP_937857.1 | 4 - 5 | 554 | 78 | |
GenBank mRNA | AK032279.1 | - | 4 - 5 | 396 | 78 |
AK047568.1 | - | 4 - 5 | 535 | 78 | |
AK122262.1 | - | 4 - 5 | 494 | 78 | |
AK138230.1 | - | 3 - 4 | 730 | 78 | |
BC072567.1 | - | 4 - 5 | 517 | 78 | |
BC117896.1 | - | 4 - 5 | 381 | 78 | |
BC139792.1 | - | 4 - 5 | 381 | 78 |