Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291076.1 | NP_001278005.1 | 5 - 6 | 601 | 67 |
NM_001291077.1 | NP_001278006.1 | 4 - 5 | 493 | 67 | |
NM_198214.3 | NP_937857.1 | 5 - 6 | 700 | 67 | |
GenBank mRNA | AK032279.1 | - | 5 - 6 | 542 | 67 |
AK047568.1 | - | 5 - 6 | 681 | 67 | |
AK122262.1 | - | 5 - 6 | 640 | 67 | |
AK138230.1 | - | 4 - 5 | 876 | 67 | |
BC072567.1 | - | 5 - 6 | 663 | 67 | |
BC096541.1 | - | 4 - 5 | 385 | 67 | |
BC117896.1 | - | 5 - 6 | 527 | 67 | |
BC117897.1 | - | 4 - 5 | 419 | 67 | |
BC139792.1 | - | 5 - 6 | 527 | 67 |