Assay Design



Gene Transcripts

Gene Symbol
Defb28
Entrez Gene ID
545475
Gene Name
defensin beta 28
Gene Aliases
Gm1635, OTTMUSG00000015852
Chromosome Location
Chr.2: 152518255 - 152521447 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037502.2 NP_001032591.1 1 - 2 112 59
GenBank mRNA AK131731.1 - 1 - 2 113 59
DQ012039.1 - 1 - 2 112 59

View Full Product Details