Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133973.2 | NP_598734.1 | 6 - 7 | 860 | 68 |
GenBank mRNA | AK156466.1 | - | 6 - 7 | 851 | 68 |
AK172465.1 | - | 6 - 7 | 851 | 68 | |
AY223866.1 | - | 6 - 7 | 887 | 68 | |
BC023120.1 | - | 6 - 7 | 837 | 68 |