Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033223.2 | NP_001028395.1 | 5 - 6 | 687 | 110 |
NM_001039354.1 | NP_001034443.1 | 5 - 6 | 729 | 110 | |
NM_001284329.1 | NP_001271258.1 | 5 - 6 | 1446 | 110 | |
GenBank mRNA | AK077191.1 | - | 5 - 6 | 1446 | 110 |
AK160671.1 | - | 5 - 6 | 636 | 110 | |
BC029721.1 | - | 5 - 6 | 635 | 110 | |
BC105965.1 | - | 4 - 5 | 758 | 110 |